Z150762
Water aspirator pump
Short swirling
About This Item
Code UNSPSC :
41100000
Nomenclature NACRES :
NB.79
Produits recommandés
Longueur
76 mm
Description générale
Operates efficiently on both high and low water pressure. Nickel-plated 3/8 in. NPTM water inlet. Ball-type, positive-acting valve in suction tube prevents backflow should pressure be reduced. Max. vacuum is 100 mm Hg (abs).
With serrated tail and side-arm.