Principaux documents
Z107042
Supelco
Flow controllers for flash-chromatography assemblies with ball joints
joint: SJ 50/30
About This Item
Code UNSPSC :
12000000
Nomenclature NACRES :
NC.05
Joint
joint: SJ50/30
Qualité
for chromatography
Technique(s)
LPLC: suitable
Adéquation
suitable for chromatography
Compatibilité
for 50/30 standard ball joint