Principaux documents
WHA68762502
Filtres seringue Whatman® GD/X
polyethersulfone membrane, pore size 0.2 μm, diam. 25 mm, non-sterile, pk of 50 ea
Synonyme(s) :
Whatman filter, Z286567, syringe filter, syringe filters
1 PKG
570,00 €
570,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 PKG
570,00 €
About This Item
Code UNSPSC :
41104922
Nomenclature NACRES :
NB.24
570,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
polyethersulfone membrane
polypropylene housing
Stérilité
non-sterile
Caractéristiques
glass fiber prefilter
Conditionnement
pk of 50 ea
Fabricant/nom de marque
Whatman 6876-2502
Whatman Article No. 28420517 (US reference)
Diamètre
25 mm
Volume
≤25 mL
Dimension de pores
0.2 μm
Raccords
female Luer inlet (lock)
male Luer outlet
Description générale
Non-sterile syringe filters with a Polyethersulfone (PES) membrane, suitable for serum, plasma and tissue culture solutions. Whatman GD/X Syringe Filters are high-viscosity filters that incorporate a prefilter to enable filtration of viscous, dense or otherwise hard-to-filter material with high solids content as in environmental and food analysis samples. The prefilter contains a high-efficiency borosilicate glass filtration stack that will retain fine particles down to 0.7 μm, allowing difficult sample filtration with less hand force. It reduces the need for multiple syringe filters by processing three to seven times more sample volume than syringe filters without prefilters. The filters can be autoclaved.
Caractéristiques et avantages
  • Non sterile Whatman GD/X Syringe Filters combine a polypropylene housing, high-efficiency Whatman GMF 150 and GF/F pre-filter media, and one of the following membrane materials:
  • Polyethersulfone (PES) for serum, plasma and tissue culture
  • Polyvinylidene difluoride (PVDF) for HPLC
  • Cellulose acetate for biological solutions
  • Borosilicate glass microfiber
Adéquation
Suitable for filtering environmental and food analysis samples. Suitable for particulate laden sample preparation.
Autres remarques
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Informations légales
Whatman is a registered trademark of Cytiva