Documents
TEM-LFH15100AR1.3
Sigma-Aldrich
QUANTIFOIL on London Finder Grids (H15)
Gold Grids, size 135 mesh, pack of 100
About This Item
Code UNSPSC :
41111761
Nomenclature NACRES :
MA.24
Conditionnement
pack of 100
Taille
135 mesh
Description générale
QUANTIFOIL Gold Grids, 200 Mesh, 1.2 micron hole, 2.5 micron period. London Finder (H15), Pitch 188 micron, 135 mesh