Principaux documents
TEM-CF300AU
Sigma-Aldrich
Carbon Film Supported Gold Square Mesh
size 300 mesh, box of 25
Synonyme(s) :
CARBON FILM 300# GOLD 25/PK
About This Item
Code UNSPSC :
41111761
Conditionnement
box of 25
Taille
300 mesh
Description générale
A 5-6 nm thin film of pure carbon supported standard squre mesh grids. Gold grid, 300 mesh.