Principaux documents
TEM-CF150CU
Sigma-Aldrich
Carbon Film Supported Copper Square Mesh
size 150 mesh, box of 25
Synonyme(s) :
CARBON FILM 150 MESH CU 25/BX
About This Item
Code UNSPSC :
41111761
Conditionnement
box of 25
Taille
150 mesh
Description générale
A 5-6 nm thin film of pure carbon supported standard squre mesh grids. Copper grid, 150 mesh.