Principaux documents
TEM-CF150AU50
Sigma-Aldrich
Carbon Film Supported Gold Square Mesh
size 150 mesh, box of 50
Synonyme(s) :
CARBON FILM 150 MESH AU 50/BX
About This Item
Code UNSPSC :
41111761
Conditionnement
box of 50
Taille
150 mesh
Description générale
A 5-6 nm thin film of pure carbon supported standard squre mesh grids. Gold grid, 150 mesh.