TEM-71165-50
Cryogenic Grid Box Handling Tool
About This Item
Code UNSPSC :
41111761
Nomenclature NACRES :
MA.24
Produits recommandés
Catégories apparentées
Description générale
This tool has one end which is threaded and fits into the center hole of the Cryogenic Grid Storage Box (where the screw goes in to secure the lid) for moving the box in and out of the cryogenic chamber.