SYNS181125
Synthware™ straight bore PTFE stopcock plug
bore size 2 mm, plug size: 11/25
Synonyme(s) :
Synthware S181125
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware S181125
Taille de l'alésage
2 mm
Description générale
This replacement kit is complete with one PTFE plug (1:5 taper), one PTFE washer, one O-ring.
Informations légales
Synthware is a trademark of Kemtech America Inc.