Principaux documents
SYNH121410
Synthware Hickman still head with side port, cap, o-ring and PTFE-faced septum
joint: ST/NS 14/10
Synonyme(s) :
Synthware H121410, Hickman Distillation head
1 EA
53,10 €
53,10 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 EA
53,10 €
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.51
53,10 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Joint
joint: ST/NS14/10
Fabricant/nom de marque
H121410
Description générale
Synthware Hickman distillation head is designed for microscale fractional distillation. It is supplied with one compression cap, one Viton® O-ring, and one PTFE-faced silicone rubber septum. The side port includes a cap and an 8 mm PTFE-lined septum for syringe injection or sample retrieval.
Informations légales
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company