SYNC184603
Synthware™ chromatography column with PTFE stopcock
top joint: ST/NS 24/40, I.D. × L 53.0 mm × 305 mm
Synonyme(s) :
Synthware C184603, glass chromatograpy column
About This Item
Produits recommandés
Joint
joint: ST/NS top24/40
Qualité
for chromatography
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware C184603
D.I. × L
53.0 mm × 305 mm
Description générale
This product does not include coarse fritted disc.