Documents
SYNA891420
Synthware distilling adapter with 105 degree bend
top joint: ST/NS 14/20, bottom joint: ST/NS 14/20
Synonyme(s) :
Synthware 891420, bent distillation adapter
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.51
Joint
bottom joint: ST/NS14/20
joint: ST/NS top14/20
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware A891420
Catégories apparentées
Description générale
With an inner drip joint at the bottom and an outer joint at the top.
Informations légales
Synthware is a trademark of Kemtech America Inc.