Principaux documents
SYNA253424
Synthware enlarging adapter
top joint: ST/NS 34/45, bottom joint: ST/NS 24/40
Synonyme(s) :
Synthware A253424, glass connecting adapter
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.51
Joint
bottom joint: ST/NS24/40
joint: ST/NS top34/45
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware A253424
Catégories apparentées
Description générale
Synthware enlarging adapter is used for connecting dissimilar standard taper joint sizes. The top joint (34/45) is larger that the bottom joint (24/40).
Informations légales
Synthware is a trademark of Kemtech America Inc.