Principaux documents
DWK92812G-200
KIMBLE® KIMAX® Heavy duty volumetric wide-mouth flask with glass stopper
borosilicate glass flask, flask capacity (200 mL), class A
6 EA
510,00 €
510,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
6 EA
510,00 €
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.43
510,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
borosilicate glass flask
glass stopper
Caractéristiques
class A
Conditionnement
case of 6 ea
Fabricant/nom de marque
Kimble® 92812G-200
Contenance du flacon/de la fiole
(200 mL)
Description générale
Class A Heavy Duty Wide Mouth Volumetric Flasks with Glass Stopper
  • Pipet access is easy with wide-mouth volumetric flasks
  • Superior strength, durability and safety from heavy, uniform walls
  • Quick identification with large, permanent, easy-to-read markings
  • Flat interior bottoms for stirring with standard stir bars
  • Graduated to Class A volumetric tolerances
  • Standard Taper glass stoppers
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Caractéristiques et avantages
  • Manufactured from borosilicate glass 3.3
  • Wide mouth opening increases efficiency when performing large numbers of dilutions using volumetric pipets
  • Heavy duty walls provide superior strength, durability and safety
  • Wide mouth minimizes glass-to-glass contact allowing greater flexibility in hand movements which reduces the risk of spills
  • Wide mouth opening increases efficiency when performing large numbers of dilutions using volumetric pipets
  • Printed large volume numbers allow quick identification when selecting glassware
  • Flat interior bottoms for stirring with standard stir bars
  • Calibrate to contain
  • Graduated to Class A volumetric tolerances
  • Supplied with or without stopper
Informations légales
KIMBLE is a registered trademark of DWK Life Sciences
Kimax is a registered trademark of Gerresheimer Glass, Inc.