Principaux documents
DWK213533306
Duran® Funnel
long stem(s)
Synonyme(s) :
213533306
About This Item
Code UNSPSC :
41121809
Nomenclature NACRES :
NB.43
Matériaux
long stem(s)
Conditionnement
pkg of 10 ea
Fabricant/nom de marque
DURAN® 213533306
Diam. de l'entonnoir
55 mm
Application
Decanting and filtration of liquids, The spillage of substances is avoided, Filtering, Boro 3.3
Caractéristiques et avantages
With long stem; Due to its shape, it is suitable for liquid-solid filtration using a filter paper; Fired-on,
Informations légales
DURAN is a registered trademark of DWK Life Sciences