Principaux documents
CLS999818
Corning® phenolic cap, PTFE liner
18-415 thread
Synonyme(s) :
black phenolic, phenolic caps, screw caps, Corning® 9998
192 EA
134,00 €
134,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
192 EA
134,00 €
About This Item
Code UNSPSC :
41122500
Nomenclature NACRES :
NB.43
134,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
PTFE liner
black phenolic cap
Fabricant/nom de marque
Corning 9998-18
Hauteur
~17.5 mm
Raccords
18-415 thread
Description générale
These reusable Corning caps are fabricated from specially formulated resin to resist the effects of temperature and steam in autoclaving. Caps are provided with a glued-in rubber liner over which is firmly bonded a PTFE interface to provide a highly inert sealing face.
Informations légales
Corning is a registered trademark of Corning, Inc.