Principaux documents
CLS564650
Pyrex® Verified volumetric flasks
volume 50 mL
Synonyme(s) :
flasks glass, volumetric flasks, volumetric glass, Corning® 5646
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.43
Le tarif et la disponibilité ne sont pas disponibles actuellement.
Matériaux
borosilicate glass
long
Stérilité
non-sterile
Caractéristiques
closure type ST stopper and Polyethylene stopper
Conditionnement
case of 12
Fabricant/nom de marque
Corning 5646-50
Capacité
50 mL
Volume
50 mL
Description générale
Each individual flask′s measurement has been validated per ASTM E-542 during production, which guarantees conformance to Class A specifications called out in ASTM E-288. Each flask is labeled with "Verified" and a lot number. With this information, you can trace your glassware, access a Certificate of Analysis with lot specific data regarding both measurements and methods, and track usage.
All flasks are supplied with both a Pyrex stopper and HDPE stopper.
Informations légales
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.