Principaux documents
CLS5563550
Pyrex® volumetric flask, low actinic, heavy duty wide mouth, with Pyrex stopper
capacity 50 mL
Synonyme(s) :
Corning® 55635
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.43
Le produit CLS5563550 n'est pas actuellement commercialisé dans votre pays Contacter notre Service technique
Taille de l'obturateur
13
Fabricant/nom de marque
Corning 55635-50
Capacité
50 mL
Description générale
Low actinic stained glass provides protection for materials sensitive to light. the protective color is an intergral part of the flask, which retains the mechanical strength, chemical stability and thermal resistance of the Pyrex brand labware.
Informations légales
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.