Principaux documents
BR782151
BRAND® lids for microplates BRAND®plates
plate size 96 wells, without condensation rings
Synonyme(s) :
Accessories, BRANDplates®, Lids, Microplates
100 EA
85,50 €
85,50 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
100 EA
85,50 €
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.15
85,50 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
polystyrene
Conditionnement
pack of 100 ea
Fabricant/nom de marque
BRAND 782151
Taille de la plaque/du plateau
96 wells
Adéquation
suitable for (bacteriological assays)
Description générale
To protect samples, reaction batches or cell cultures from both contamination and evaporation, compatible lids are available for all BRANDplates® microplates. The lids are made of crystal-clear polystyrene. They are optimally matched to the corresponding BRANDplates® microplates, so that plates with lids can also be stacked. The cut-corner sets the orientation of the lid. For 96-well standard plates, lids with condensation rings are available.
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG
BRANDplates is a registered trademark of BRAND GMBH + CO KG