Principaux documents
BR709133
BRAND® disposable BLAUBRAND® micropipettes, intraEND
tube capacity 50 μL
Synonyme(s) :
Micropipettes, Pipettes, intraEND
About This Item
Code UNSPSC :
41121500
Nomenclature NACRES :
NB.01
Caractéristiques
DE-M marking, To Contain
Conditionnement
pack of 1000 ea (4 boxes of 250)
Fabricant/nom de marque
BRAND 709133
Longueur
32 mm
Capacité du tube
50 μL
Description générale
DE-M marking certified for volumes greater than 5 μl. Volume defined by end-to-end filling. Calibrated to contain (TC, In).
Informations légales
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG