BR65231
BRAND® desiccator ′Novus′
nominal diameter × O.D. × H 150 mm × 215 mm × 316 mm, top socket joint: 24/29
About This Item
Code UNSPSC :
41104510
Nomenclature NACRES :
NB.43
Produits recommandés
Joint
socket joint: top24/29
Fabricant/nom de marque
BRAND 65231
Diamètre nominal × D.E. × H
150 mm × 215 mm × 316 mm
Catégories apparentées
Description générale
Boro 3.3. DIN 12491. Precisely ground flat flanges. Base ring surface studded to improve stability and help prevent cracks. Interchangeable lid. Desiccators with sockets are supplied with 24/29 socket, and interchangeable Boro 3.3 stopcock.
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG