Documents
BR362338
BRAND® BLAUBRAND® ASTM 100 mL centrifuge tube
cylindrical, conical bottom, former standard ASTM D96
Synonyme(s) :
ASTM, BLAUBRAND®, Centrifuge Tubes, Tubes
About This Item
Code UNSPSC :
41121703
Nomenclature NACRES :
NB.11
Matériaux
borosilicate glass
conical bottom
cylindrical
Stérilité
non-sterile
Caractéristiques
RCF 700 × g
Conditionnement
pack of 2
Fabricant/nom de marque
BRAND 362338
Longueur
167 mm
Description générale
Withstands RCF up to 700. Design, accuracy, etc. to meet ASTM ("American Society for Testing and Materials") specifications.
Informations légales
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG