BR36172
BRAND® volumetric flask
PMP, transparent, size 25 mL
Synonyme(s) :
Plastics, Volumetric flasks
12,90 €
About This Item
12,90 €
Produits recommandés
Matériaux
PMP
polypropylene stopper
transparent
Joint
joint ST/NS10/19
Caractéristiques
accuracy class A
Conditionnement
pack of 1 ea
Fabricant/nom de marque
BRAND 36172
Taille
25 mL
Catégories apparentées
Description générale
Calibrated to contain (TC, In). Exposure to temperatures up to 121 °C (autoclaving) will not cause permanently exceeded error limits!
Cleaning up to max. 60 °C is recommended to preserve marks and inscriptions.