Principaux documents
BR150920
BRAND® watch glass
PTFE, diam. 125 mm
Synonyme(s) :
Dishes, PTFE, Watch glasses
1 EA
37,00 €
37,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 EA
37,00 €
About This Item
Code UNSPSC :
41121801
Nomenclature NACRES :
NB.43
37,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
PTFE
Fabricant/nom de marque
BRAND 150920
Diamètre
125 mm
Catégories apparentées
Description générale
The BRAND® watch glass is made from high-quality PTFE, making it ideal for covering PTFE beakers. Its excellent chemical resistance ensures durability and reliability when handling a wide range of substances. This watch glass provides a secure cover to prevent contamination and evaporation during experiments. Its lightweight and non-reactive properties make it a valuable accessory in laboratory settings, enhancing the efficiency of various applications. This versatile tool is essential for maintaining the integrity of samples and solutions in the lab.
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG