Principaux documents
BAF378980000
SP Bel-Art® Pipette Pump
capacity 10 mL, green
Synonyme(s) :
Pump For Pipette, pipetting device
1 EACH
39,30 €
39,30 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 EACH
39,30 €
About This Item
Code UNSPSC :
41122800
Nomenclature NACRES :
NB.01
39,30 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
green
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Bel-Art F37898-0000
Capacité
10 mL
Description générale
Provides safe, accurate and trouble-free pipetting with simple, one-handed operation.
Caractéristiques et avantages
  • Pipettes fit smoothly into its flexible tapered chuck
  • Thumbwheel on side is rotated for precision aspirating or dispensing
  • Plunger may be pressed for quick emptying
  • Resistant to acids and alkalis
  • Disassemble all parts for cleaning and before autoclaving at 121ºC (250ºF) for 20 minutes; chuck should not be autoclaved
  • Replacement Silicone Rubber Chuck and Washers are available
Informations légales
Bel-Art is a registered trademark of SP Scienceware, a division of SP