BAF371220020
SP Bel-Art® Spinbar® magnetic stir bar
polygon white PTFE-coated, L × diam. 0.75 in. (20 mm) × 0.25 in. (6 mm)
Synonyme(s) :
Spinbar® magnetic stir bar
11,20 €
About This Item
11,20 €
Produits recommandés
Matériaux
polygon white PTFE-coated
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Bel-Art F37122-0020
L × diam.
0.75 in. (20 mm) × 0.25 in. (6 mm)
Catégories apparentées
Description générale
- Spinbar® magnetic stir bar is used in conjunction with a magnetic stirring plate for uniform, consistent, and thorough mixing of chemical solutions.
- Alnico magnet encapsulated in durable and inert FDA grade polytetrafluoroethylene (PTFE) low friction coating.
- Multi-faceted-shaped bars provide a greater surface area and added turbulence for more vigorous, thorough mixing.
- Autoclave safe.
Caractéristiques et avantages
- Center molded pivot ring minimizes the contact area between the bar and the vessel; reduces friction and chattering
- Marring of plastic containers is diminished
- Alnico magnet encapsulated in FDA Grade, USP Class VI PTFE coating – Superior Chemical Compatibility
- Manufactured in a registered ISO 9001:2015 facility, as verified by SGS certification
- Autoclave safe