BAF371090032
SP Bel-Art® Spinbar® magnetic stir bar
octagonal yellow PTFE-coated, L × diam. 3 in. (76.2 mm) × 0.5 in. (12.7 mm)
Synonyme(s) :
Spinbar® magnetic stir bar
46,10 €
About This Item
46,10 €
Produits recommandés
Matériaux
octagonal yellow PTFE-coated
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Bel-Art F37109-0032
L × diam.
3 in. (76.2 mm) × 0.5 in. (12.7 mm)
Catégories apparentées
Description générale
- Spinbar® magnetic stir bar is used in conjunction with a magnetic stirring plate for uniform, consistent, and thorough mixing of chemical solutions.
- Alnico magnet encapsulated in durable and inert FDA grade polytetrafluoroethylene (PTFE) low friction coating.
- Multi-faceted-shaped bars provide a greater surface area and added turbulence for more vigorous, thorough mixing.
- Autoclave safe.
Caractéristiques et avantages
- Alnico magnet encapsulated in FDA Grade, USP Class VI PTFE brand PTFE Coating – Superior Chemical Compatibility
- Interrupted profile provides greater surface area and added turbulence when compared to the smooth surface of cylindrical bars
- Pivot ring aids in reducing friction and chattering
- Autoclavable
- Manufactured in a registered ISO 9001:2015 facility, as verified by SGS certification