Principaux documents
BAF203071825
SP Bel-Art® Safe-Lab Therm-O-Vac Joint Adapter
for for 24/40 Tapered Joints
Synonyme(s) :
thermovac adapter
About This Item
Code UNSPSC :
41122800
Nomenclature NACRES :
NB.43
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Bel-Art F20307-1825
Diam. des orifices
6 mm
Raccords
for for 24/40 Tapered Joints
Description générale
PTFE adapter with Viton® O-ring on the body, screw cap and hose connection.
Caractéristiques et avantages
Not autoclavable
Informations légales
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Viton is a registered trademark of E. I. du Pont de Nemours and Company