Principaux documents
BAB602102800
SP Bel-Art® H-B Frio-Temp® Calibrated Electronic Verification Thermometer
temp. range at -50-300 °C, Freezer Calibration
Synonyme(s) :
Calibrated freezer thermometer
About This Item
Code UNSPSC :
41112303
Nomenclature NACRES :
NB.71
Matériaux
glass bottle
stainless steel probe
Caractéristiques
Alarm, Timer
Freezer Calibration
Conditionnement
pack of 1 ea
Fabricant/nom de marque
HB Instruments B60210-2800
range of application measuring range
-50-300 °C, resolution: 1 °F/C (±1°C from -20/130°C; ±2°C from -50/-21°C and 131/200°C; otherwise ±4°C)
Paramètres
temp. range at -50-300 °C
H × l
18 mm × 52 mm
L de la sonde
105 mm , 4.1"
Alimentation par piles
AAA
Description générale
Digital thermometers verify sample temperatures in freezers, refrigerators, incubators and ovens. Available calibrated for specific applications and equipment
Note: Plastic bottles as indicated in table cannot be used in ovens. Glass bottle and cap included with products as indicated in the table should not be used in conditions where temperatures may exceed 200°C.
Caractéristiques et avantages
  • Accurate temperature reading without sample contamination is achieved by using the filled bottle (included), which mimics the sample
  • All models include accredited calibration documentation that meets the requirements of ISO 17025
  • Tested and calibrated in H-B’s exclusive triple accredited/registered ISO/IEC 17025:2017, A2LA accredited laboratory (Cert #2448.01), against equipment whose calibration is traceable to NIST
  • Includes multi-language instructions (English, French, German, Italian, Portuguese, and Spanish) Individually serialized, CE marked
Informations légales
Bel-Art is a registered trademark of SP Scienceware, a division of SP
FRIO-Temp is a registered trademark of SP Scienceware, a division of SP