Merck
HomePolymer Synthesis TechniquesInitiator/Stabilizer FAQs
Initiator/Stabilizer FAQs
Dr. S. S. Newaz
Polyorganix Inc. Houston, TX
Material Matters 2006, 1.1, 3.
Q. How does one choose an appropriate initiator?
In a free-radical addition polymerization, the choice of polymerization initiator depends mainly on two factors: a) its solubility and b) its decomposition temperature. If the polymerization is performed in an organic solvent, then the initiator should be soluble in that solvent, and the decomposition temperature of the initiator must be at or below the boiling point of the solvent. Commonly, AIBN (2,29-Azobis(2-methylpropionitrile)) (441090, dec.102–104 °C) and BPO (2-(4-Biphenyl)-5-phenyloxazole) (216984, mp 115–119 °C) suit these requirements. If the desired polymerization occurs at or below 20 °C, then special, low-temperature free-radical initiators need to be used. Various azo-type initiators can be chosen to satisfy the decomposition temperature requirement.
For emulsion polymerization or polymerization in an aqueous system, a water-soluble initiator like K2S2O8 (379824) or an organic, water-soluble initiator (4,49-Azobis(4-cyanovaleric acid)) (118168, dec.118–125 °C) would be suitable.
Determination of monomer reactivity is not always obvious or straightforward. Researchers rely on their experience and published data on individual monomers. In general, extent of conjugation in the molecular structure can be viewed as indicative of its tendency to form the initial free radical required for propagating a freeradical polymerization. Usually, a more conjugated system is more likely to undergo free-radical polymerization.
To inhibit polymerization during storage, many monomers are provided with a stabilizer as indicated by the label. Usually, it is not necessary to remove stabilizers. They are typically present in ppm level, and the use of a free radical initiator at the polymerization temperature will overwhelm the effect of the stabilizer. In worstcase scenarios, one may need to add extra amounts of initiator to sustain an acceptable polymerization rate. In most cases, once a polymerization initiates, the rate can be sustained without much difficulty. If, however, it is absolutely necessary to remove the stabilizer, column chromatography is the preferred method (for inhibitor removal columns, see products 306312311332306320).
It is a common practice to dissolve the polymers in a solvent prior to end use, followed by precipitating the polymer using a cosolvent. Usually, the residual initiators and stabilizers will remain in solution and the polymers will separate out as a solid (powder, gum, or fibers). This process may be repeated until desirable polymer characteristics are obtained. This fractional precipitation is also effective in removing lower molecular weight polymers, resulting in narrower molecular weight distribution—of course accompanied with a loss of yield. Typical solvent/cosolvent pairs could be toluene/hexane, toluene/methanol, THF/water, etc., determined by the relative solubilities of the polymer versus the small-molecule component.
For initiator solubilities and decomposition temperatures, visit our website.