Skip to Content
Merck
All Photos(2)

Key Documents

216984

Sigma-Aldrich

2-(4-Biphenyl)-5-phenyloxazole

98%

Synonym(s):

BPO

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H15NO
CAS Number:
Molecular Weight:
297.35
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

98%

mp

115-119 °C (lit.)

storage temp.

−20°C

SMILES string

c1ccc(cc1)-c2ccc(cc2)-c3ncc(o3)-c4ccccc4

InChI

1S/C21H15NO/c1-3-7-16(8-4-1)17-11-13-19(14-12-17)21-22-15-20(23-21)18-9-5-2-6-10-18/h1-15H

InChI key

RQGVWYIZAUHBBE-UHFFFAOYSA-N

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 4 - Eye Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Susy Lopes et al.
The journal of physical chemistry. A, 114(34), 9074-9082 (2010-08-10)
Methyl 4-chloro-5-phenyl-1,3-oxazole-2-carboxylate (MCPOC) has been synthesized and isolated in cryogenic matrices (argon and xenon). FTIR spectroscopy studies on the matrix isolated compound, supported by DFT(B3LYP)/6-311++G(d,p) calculations, allow for the identification of two low-energy conformers (I and II) of the molecule
Anna Krzywicka et al.
Nanomaterials (Basel, Switzerland), 10(11) (2020-11-18)
In the present work, we report the studies on perfectly homogeneous nanocomposites composed of polystyrene-grafted silver nanoparticles (Ag@PS) as a bioactive fulfilment and a mixture of polystyrene (PS) and polyvinyl alcohol (PVA) as a matrix. The procedure developed by our
Xiaofei Guo et al.
Dalton transactions (Cambridge, England : 2003), 47(35), 12130-12137 (2018-08-04)
A composite photocatalyst, with a BiOI/TiO2 heterojunction supported on carboxyl polystyrene spheres, was prepared using a two step hydrothermal reaction method. The products were characterized by X-ray diffraction, scanning electron microscopy and energy-dispersive X-ray spectroscopy. The results show that the
Pavlopoulos, T. G., & Hammond, P. R.
Spectroscopic studies of some laser dyes.
Journal of the American Chemical Society, 96(21), 6568-6579 (1974)
Jiaying Li et al.
International journal of molecular sciences, 20(9) (2019-04-28)
Poly(methyl methacrylate) (PMMA) bone cements have been widely used in clinical practices. In order to enhance PMMA's imaging performance to facilitate surgical procedures, a supplementation of radiopaque agent is needed. However, PMMA bone cements are still facing problems of loosening

Articles

An article regarding common FAQs for initiators and stabilizers.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service