Merck
  • Degradation of regulator of calcineurin 1 (RCAN1) is mediated by both chaperone-mediated autophagy and ubiquitin proteasome pathways.
Degradation of regulator of calcineurin 1 (RCAN1) is mediated by both chaperone-mediated autophagy and ubiquitin proteasome pathways.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2009-06-11)
Heng Liu, Pin Wang, Weihong Song, Xiulian Sun
ABSTRACT
Regulator of calcineurin 1 (RCAN1), a gene identified from the critical region of Down syndrome, has been implied in pathogenesis of Alzheimer's disease (AD). RCAN1 expression was shown to be increased in AD brains; however, the mechanism of RCAN1 gene regulation is not well defined. The present study was designed to investigate the molecular mechanism of RCAN1 protein degradation. In addition to being degraded through the ubiquitin proteasome pathway, we found that lysosomal inhibition markedly increased RCAN1 protein expression in a time- and dosage-dependent manner. Inhibition of macroautophagy reduced RCAN1 expression, indicating that RCAN1 degradation is not through a macroautophagy pathway. However, disruption of chaperone-mediated autophagy (CMA) increased RCAN1 expression. Two CMA recognition motifs were identified in RCAN1 protein to mediate its degradation through a CMA-lysosome pathway. A promoter assay further demonstrated that inhibition of RCAN1 degradation in cells reduced calcineurin-NFAT activity. Dysfunctions of ubiquitin-proteasome and autophagy-lysosome pathways have been implicated in neurodegenerative diseases. Therefore, elucidation of RCAN1 degradation by a ubiquitin proteasome pathway and CMA-lysosome pathway in the present study may greatly advance our understanding of AD pathogenesis.
Sigma-Aldrich
Chloroquine diphosphate salt, powder or crystals, 98.5-101.0% (EP)
Sigma-Aldrich
3-Methyladenine, autophagy inhibitor
Roche
cOmplete Protease Inhibitor Cocktail, Tablets provided in EASYpacks
Sigma-Aldrich
Rapamycin from Streptomyces hygroscopicus, ≥95% (HPLC), powder
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Lactacystin, Synthetic, Lactacystin, Synthetic, CAS 1258004-00-0, is an irreversible inhibitor of 20S proteasome inhibitor (IC₅₀ = 500 nM). A covalent inhibitor of the chymotrypsin & trypsin-like activities of proteasome.