Skip to Content
Merck
All Photos(1)

Documents

426100

Sigma-Aldrich

Lactacystin

≥99% (HPLC), solid, proteasome inhibitor, Calbiochem®

Synonym(s):

Lactacystin, Synthetic, Proteasome Inhibitor VI

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H24N2O7S
CAS Number:
Molecular Weight:
376.43
UNSPSC Code:
12352200
NACRES:
NA.77

product name

Lactacystin, Synthetic, Lactacystin, Synthetic, CAS 1258004-00-0, is an irreversible inhibitor of 20S proteasome inhibitor (IC₅₀ = 500 nM). A covalent inhibitor of the chymotrypsin & trypsin-like activities of proteasome.

Quality Level

Assay

≥99% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

DMSO: 50 mg/mL
water: soluble

shipped in

ambient

storage temp.

−20°C

General description

An irreversible proteasome inhibitor. A Streptomyces metabolite that acts as a highly specific inhibitor of the 20S proteasome (MCP; multicatalytic proteinase complex) (IC50 = 500 nM for 20S proteasome). Blocks proteasome activity by targeting the catalytic β-subunit. Acts as a covalent inhibitor of the chymotrypsin- and trypsin-like activities of the proteasome. Induces neurite outgrowth in Neuro 2A mouse neuroblastoma cells and inhibits progression of synchronized Neuro 2A cells and MG-63 human osteosarcoma cells beyond the G1 phase of the cell cycle. Reported to induce apoptosis in human monoblast U937 cells. Also inhibits NF-κB activation (IC50 = 10 µM).
An irreversible proteasome inhibitor. A Streptomyces metabolite that acts as a highly specific inhibitor of the 20S proteasome (MCP; multicatalytic proteinase complex) (IC50 = 500 nM for 20S proteasome). Blocks proteasome activity by targeting the catalytic β-subunit. Acts as a covalent inhibitor of the chymotrypsin and trypsin-like activities of the proteasome. Induces neurite outgrowth in Neuro 2A mouse neuroblastoma cells beyond theG1 phase of the cell cycle. Reported to induce apoptosis in human monoblast U937 cells. Also inhibits NF-κB activation (IC50 = 10 µM)

Biochem/physiol Actions

Cell permeable: no
Primary Target
20S proteasome (MCP; multicatalytic proteinase complex)
Product does not compete with ATP.
Reversible: no
Target IC50: 500 nM against 20S proteasome; 10 µM against NF-κB activation

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution in DMSO, aliquot and freeze (-20°C). DMSO stock solutions are stable for up to 1 month at -20°C. Unstable in aqueous solutions; reconstitute in H₂O just prior to use.

Other Notes

Keyomarsi, K., et al. 2011. Cell. Biol. Toxicol.27, 123.
Adams, J., and Stein, R. 1996. Ann. Rep. Med. Chem.31, 279.
Dick, L.R., et al. 1996. J. Biol. Chem.271, 7273.
Oda, K., et al. 1996. Biochem. Biophys. Res.Commun.219, 800.
Fenteany, G., et al. 1995. Science268, 726.
Imajoh-Ohmi, S., et al. 1995. Biochem.Biophys. Res. Commun.217, 1070.
Jensen, T.J., et al. 1995. Cell 83, 129.
Katagiri, M.M., et al. 1995. J. Antibiot.48, 344.
Mori, S., et al. 1995. J. Biol. Chem.270, 29447.
Tanaka, H., et al. 1995. Biochem. Biophys. Res. Commun.216, 291.
Fenteany, G., et al. 1994. Proc. Natl. Acad.Sci. USA91, 3358.
Omura, S., et al. 1991. J. Antibiot.44, 117.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yue Qu et al.
Microbiology spectrum, 11(3), e0037823-e0037823 (2023-04-07)
Several viruses have been shown to modulate the transcription factor nuclear factor erythroid 2-related factor 2 (NRF2), the master regulator of redox homeostasis. The severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2), responsible for the COVID-19 pandemic, also seems to disrupt
Andrew M Pickering et al.
The Biochemical journal, 432(3), 585-594 (2010-10-06)
Oxidized cytoplasmic and nuclear proteins are normally degraded by the proteasome, but accumulate with age and disease. We demonstrate the importance of various forms of the proteasome during transient (reversible) adaptation (hormesis), to oxidative stress in murine embryonic fibroblasts. Adaptation
Yiming Lei et al.
Autophagy, 17(12), 4423-4441 (2021-04-20)
The hepatitis B virus X protein (HBx) is involved in the process of hepatocellular carcinoma via the activation of various oncogenes. Our previous study indicated that ARBB1 (arrestin beta 1) promotes hepatocellular carcinogenesis (HCC). However, the role of ARRB1 in
Kazuhiko Watabe et al.
Neuropathology : official journal of the Japanese Society of Neuropathology, 42(6), 488-504 (2022-06-16)
The formation of misfolded protein aggregates is one of the pathological hallmarks of neurodegenerative diseases. We have previously demonstrated the cytoplasmic aggregate formation of adenovirally expressed transactivation response DNA-binding protein of 43 kDa (TDP-43), the main constituent of neuronal cytoplasmic aggregates
Diti Chatterjee Bhowmick et al.
The Journal of biological chemistry, 293(37), 14210-14223 (2018-07-18)
Human islet amyloid polypeptide (hIAPP) is the principal constituent of amyloid deposits and toxic oligomers in the pancreatic islets. Together with hyperglycemia, hIAPP-derived oligomers and aggregates are important culprits in type 2 diabetes mellitus (T2DM). Here, we explored the role

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service