Merck
  • Tumor necrosis factor-alpha induces caspase-mediated cleavage of peroxisome proliferator-activated receptor gamma in adipocytes.
Tumor necrosis factor-alpha induces caspase-mediated cleavage of peroxisome proliferator-activated receptor gamma in adipocytes.
The Journal of biological chemistry (2009-03-27)
Adilson Guilherme, Gregory J Tesz, Kalyani V P Guntur, Michael P Czech
ABSTRACT
The nuclear receptor peroxisome proliferator-activated receptor gamma (PPARgamma) is a ligand-dependent transcription factor that acts as a primary regulator of adipogenesis and controls adipocyte metabolism and insulin action. Increased expression of tumor necrosis factor (TNFalpha) in adipose tissue of obese subjects potently suppresses the expression of PPARgamma and attenuates adipocyte functions. Here we show that PPARgamma is a substrate of caspase-3 and caspase-6 during TNFalpha receptor signaling in adipocytes, and the consequent PPARgamma cleavage disrupts its nuclear localization. TNFalpha treatment of 3T3-L1 adipocytes decreases full-length PPARgamma while increasing the level of a 45-kDa immunoreactive PPARgamma fragment. Specific inhibitors of caspase-3 and caspase-6 attenuate the cleavage of PPARgamma protein in response to TNFalpha in cultured adipocytes. Incubation of nuclear fractions with recombinant caspase-3 and caspase-6 also generates a 45-kDa PPARgamma cleavage product. Dispersion of nuclear PPARgamma to the cytoplasm in response to TNFalpha treatment occurs in parallel with detection of activated caspase-3. We suggest that activation of the caspase cascade by TNFalpha down-regulates PPARgamma protein and PPARgamma-mediated metabolic processes in adipose cells.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Lactacystin, Synthetic, Lactacystin, Synthetic, CAS 1258004-00-0, is an irreversible inhibitor of 20S proteasome inhibitor (IC₅₀ = 500 nM). A covalent inhibitor of the chymotrypsin & trypsin-like activities of proteasome.
Sigma-Aldrich
TNF-α, Mouse, Recombinant, E. coli
Sigma-Aldrich
Caspase Inhibitor III, The Caspase Inhibitor III, also referenced under CAS 634911-80-1, controls the biological activity of Caspase. This small molecule/inhibitor is primarily used for Cancer applications.
Sigma-Aldrich
MG-132, InSolution, ≥98%, 10 mM, reversible proteasome inhibitor