Merck
  • Surface characterization and drug release from porous microparticles based on methacrylic monomers and xanthan.
Surface characterization and drug release from porous microparticles based on methacrylic monomers and xanthan.
Carbohydrate polymers (2015-04-11)
Maria-Andreea Lungan, Marcel Popa, Stefania Racovita, Gabriela Hitruc, Florica Doroftei, Jacques Desbrieres, Silvia Vasiliu
ABSTRACT
Porous crosslinked microparticles based on glycidyl methacrylate and xanthan were prepared by suspension polymerization and used for loading theophylline, a bronhodilatator drug, in order to obtain new drug delivery systems. The surface morphologies observed by means of SEM and AFM analysis demonstrated that microparticles show a spherical shape and are characterized by a porous structure. The presence of xanthan in the structure of microparticles leads to a decrease of surface roughness and pore diameters as well as to an increase of hydrophilicity degree compared to the micropaticles based only on glycidyl methacrylate. To analyze the in vitro release data various mathematical models were used, such as, first order, Higuchi model, Korsmeyer-Peppas model and Baker-Lonsdale model. Based on the highest values of the correlation coefficient, the analysis of the kinetic data indicate that drug release from G1 and X1 porous microparticles fits similarly well to the first order and Higuchi models and diffusion was the dominant mechanism of drug release.
Sigma-Aldrich
Triethylene glycol dimethacrylate, contains 200 ppm monomethyl ether hydroquinone as inhibitor, 99%
Sigma-Aldrich
Adenosine 5′-phosphosulfate sodium salt, ≥85%
Sigma-Aldrich
Benzoyl peroxide blend with dicyclohexyl phthalate, suitable for use as a catalyst for electron microscopy. Modified to render it safe in transit.
Sigma-Aldrich
Ethylene glycol dimethacrylate, 98%, contains 90-110 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
(Benzotriazol-1-yloxy)tris(dimethylamino)phosphonium hexafluorophosphate, 97%
Sigma-Aldrich
Luperox® A75FP, Benzoyl peroxide, 75% remainder water, contains 25 wt. % water as stabilizer, 75%
Sigma-Aldrich
Luperox® A75, Benzoyl peroxide, 75%, remainder water
Sigma-Aldrich
Glycidyl methacrylate, 97%, contains 100 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
Triethylene glycol dimethacrylate, contains 80-120 ppm MEHQ as inhibitor, 95%
Sigma-Aldrich
Theophylline, anhydrous, ≥99%, powder
Sigma-Aldrich
Glycidyl methacrylate, ≥97.0% (GC)
Sigma-Aldrich
Ammonium persulfate, BioXtra, ≥98.0%
Sigma-Aldrich
Ammonium persulfate, BioUltra, for molecular biology, ≥98.0% (RT)
Sigma-Aldrich
Ammonium persulfate, for molecular biology, suitable for electrophoresis, ≥98%