Skip to Content
Merck
All Photos(4)

Key Documents

517909

Sigma-Aldrich

Luperox® A75, Benzoyl peroxide

75%, remainder water

Synonym(s):

Benzoyl peroxide, Dibenzoyl peroxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5CO)2O2
CAS Number:
Molecular Weight:
242.23
Beilstein:
984320
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

75%

autoignition temp.

176 °F

reaction suitability

reagent type: oxidant

mp

105 °C (lit.)

storage temp.

2-8°C

SMILES string

O=C(OOC(=O)c1ccccc1)c2ccccc2

InChI

1S/C14H10O4/c15-13(11-7-3-1-4-8-11)17-18-14(16)12-9-5-2-6-10-12/h1-10H

InChI key

OMPJBNCRMGITSC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Benzoyl peroxide belongs to a family of peroxides. It consists of two benzoyl groups bridged by peroxide link.Benzoyl peroxide has low solubility in water and ethanol.

Application

Benzoyl peroxide can be used as:
  • An oxidizing agent in the synthesis of conducting polypyrrole via inverted emulsion polymerization.
  • An mild oxidizing agent and carboxylate source in the synthesis of substituted imides.
  • A radical initiator in the preparation of PCL/CNF:MX (poly(ε-caprolactone)/cellulose nanofibrils: compatibilizer) bio-nanocomposites.
Widely used initiator, curing agent, and cross-linking agent in polymerization processes.

Legal Information

Product of Arkema Inc.
Luperox is a registered trademark of Arkema Inc.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Org. Perox. B - Skin Sens. 1

Storage Class Code

4.1A - Other explosive hazardous materials

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of adapalene 0.1%/benzoyl peroxide 2.5% topical gel on quality of life and treatment adherence during long-term application in patients with predominantly moderate acne with or without concomitant medication-additional results from the non-interventional cohort study ELANG.
Gollnick HPM, et al.
Journal of the European Academy of Dermatology and Venereology : JEADV, 24(S4), 23-29 (2015)
Development of a sensitive long path absorption photometer to quantify peroxides in aerosol particles (Peroxide-LOPAP)."
Mertes P, et al.
Atmospheric Measurement Techniques, 5(10), 2339-2348 (2012)
Tahani Kaldéus et al.
ACS nano, 13(6), 6409-6420 (2019-05-16)
A molecularly engineered water-borne reactive compatibilizer is designed for tuning of the interface in melt-processed thermoplastic poly(caprolactone) (PCL)-cellulose nanocomposites. The mechanical properties of the nanocomposites are studied by tensile testing and dynamic mechanical analysis. The reactive compatibilizer is a statistical
Clindamycin phosphate 1? 2%-benzoyl peroxide 3? 0% fixed-dose combination gel has an effective and acceptable safety and tolerability profile for the treatment of acne vulgaris in Japanese patients: a phase III, multicentre, randomised, single-blinded, active-controlled, parallel-group study."
Kawashima M, et al.
British Journal of Dermatology, 172(2), 494-503 (2015)
Tobias Brandhofer et al.
The Journal of organic chemistry, 84(20), 12992-13002 (2019-08-29)
A simple and mild Cu-catalyzed oxidative three-component oxidative Ugi-type method for the synthesis of a variety of substituted imides has been developed. In this direct imidation approach, benzoyl peroxide serves as both the oxidant and the carboxylate source, allowing not

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service