Merck
  • MET receptor is a potential therapeutic target in high grade cervical cancer.
MET receptor is a potential therapeutic target in high grade cervical cancer.
Oncotarget (2015-04-19)
Katarzyna Miekus, Marta Pawlowska, Małgorzata Sekuła, Grazyna Drabik, Zbigniew Madeja, Dariusz Adamek, Marcin Majka
ABSTRACT
Cervical cancer is one of the leading causes of death among women suffering from tumors. Current treatment options are insufficient. Here, we investigated the MET receptor as a potential molecular target in advanced cervical cancer. Downregulation of MET receptor expression via RNA interference in different cervical carcinoma cell lines dramatically decreased tumor growth and forced tumor differentiation in vivo. MET receptor silencing also led to a dramatic decrease in cell size and a decrease in proliferation rate under normal and stress conditions. MET receptor downregulation also resulted in decreased cyclin D1 and c-myc levels but did not increase apoptosis. Subsequent experiments showed that downregulation of the MET receptor decreased the expression of a key regulator of the epithelial-to-mesenchymal transition, SLUG. and increased the expression of E-cadherin, a hallmark of the epithelial phenotype. Moreover, MET downregulation impairs expression and signaling of CXCR4 receptor, responsible for invasive phenotype. Taken together, our results strongly suggest that the MET receptor influences the oncogenic properties of cervical carcinoma cells in vitro and in vivo. These findings highlight a unique role of the MET receptor in cervical carcinoma cells and indicate the MET receptor as a potential therapeutic target for advanced cervical carcinoma.
Sigma-Aldrich
MISSION® esiRNA, targeting human MET
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Rnmt
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Phenylacetic acid, 99%
Sigma-Aldrich
Phenylacetic acid, ≥99%, FCC, FG
Sigma-Aldrich
MISSION® esiRNA, targeting human RNMT
Sigma-Aldrich
Citrate Concentrated Solution, BioUltra, for molecular biology, 1 M in H2O
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%