Skip to Content
Merck
All Photos(1)

Key Documents

S5770

Sigma-Aldrich

Citrate Concentrated Solution

BioReagent, suitable for coagulation assays, 4 % (w/v)

Synonym(s):

Sodium citrate solution

Sign Into View Organizational & Contract Pricing

Select a Size

50 ML
€35.70

€35.70


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
50 ML
€35.70

About This Item

Empirical Formula (Hill Notation):
C6H5Na3O7
CAS Number:
Molecular Weight:
258.07
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

€35.70


Please contact Customer Service for Availability

Request a Bulk Order

sterility

aseptically filled

Quality Level

product line

BioReagent

form

liquid

concentration

4 % (w/v)

suitability

suitable for coagulation assays

storage temp.

2-8°C

SMILES string

[Na+].[Na+].[Na+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O

InChI

1S/C6H8O7.3Na/c7-3(8)1-6(13,5(11)12)2-4(9)10;;;/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);;;/q;3*+1/p-3

InChI key

HRXKRNGNAMMEHJ-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

Citrate Concentrated Solution has been used to prevent coagulation of the blood sample used for hemograms.[1] It has also been used to analyze the effect of its anticoagulant action on clumping, using platelet-rich plasma (PRP).[2]

Biochem/physiol Actions

Citrate Concentrated Solution or sodium citrate is an anticoagulant used for collecting blood samples. It is useful in coagulation testing and for the erythrocyte sedimentation rate.[3] Sodium citrate eliminates calcium, which is known to mediate coagulation.[4]

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Febrile, Allergic, and Other Noninfectious Transfusion Reactions
Blood Banking and Transfusion Medicine, 677-690 (2007)
Collection and Handling of Blood
Dacie and Lewis Practical Haematology, 1-7 (2017)
Dong Hyun Jo et al.
Oncotarget, 5(22), 11513-11525 (2014-11-02)
Retinoblastoma, the most common intraocular malignant tumor in children, is characterized by the loss of both functional alleles of RB1 gene, which however alone cannot maintain malignant characteristics of retinoblastoma cells. Nevertheless, the investigation of other molecular aberrations such as
Dhiraj Kumar et al.
PloS one, 9(10), e110482-e110482 (2014-10-21)
Bisphenol-A (BPA), a well known endocrine disruptor, impairs learning and memory in rodents. However, the underlying molecular mechanism of BPA induced impairment in learning and memory is not well known. As synaptic plasticity is the cellular basis of memory, the
Yosuke Funato et al.
The Journal of clinical investigation, 124(12), 5398-5410 (2014-10-28)
Intracellular Mg(2+) levels are strictly regulated; however, the biological importance of intracellular Mg(2+) levels and the pathways that regulate them remain poorly understood. Here, we determined that intracellular Mg(2+) is important in regulating both energy metabolism and tumor progression. We

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service