Merck
  • Effects of exogenous glucagon-like peptide-1 on blood pressure, heart rate, gastric emptying, mesenteric blood flow and glycaemic responses to oral glucose in older individuals with normal glucose tolerance or type 2 diabetes.
Effects of exogenous glucagon-like peptide-1 on blood pressure, heart rate, gastric emptying, mesenteric blood flow and glycaemic responses to oral glucose in older individuals with normal glucose tolerance or type 2 diabetes.
Diabetologia (2015-06-07)
Laurence G Trahair, Michael Horowitz, Julie E Stevens, Christine Feinle-Bisset, Scott Standfield, Diana Piscitelli, Christopher K Rayner, Adam M Deane, Karen L Jones
ABSTRACT
A postprandial fall in BP occurs frequently in older individuals and in patients with type 2 diabetes. The magnitude of this decrease in BP is related to the rate of gastric emptying (GE). Intravenous administration of glucagon-like peptide-1 (GLP-1) attenuates the hypotensive response to intraduodenal glucose in healthy older individuals. We sought to determine the effects of exogenous GLP-1 on BP, GE, superior mesenteric artery (SMA) flow and glycaemic response to oral ingestion of glucose in healthy older individuals and patients with type 2 diabetes. Fourteen older volunteers (six men, eight women; age 72.1 ± 1.1 years) and ten patients with type 2 diabetes (six men, four women; age 68.7 ± 3.4 years; HbA1c 6.6 ± 0.2% [48.5 ± 2.0 mmol/mol]; nine with blood glucose managed with metformin, two with a sulfonylurea and one with a dipeptidyl-peptidase 4 inhibitor) received an i.v. infusion of GLP-1 (0.9 pmol kg(-1) min(-1)) or saline (154 mmol/l NaCl) for 150 min (t = -30 min to t = 120 min) in randomised order. At t = 0 min, volunteers consumed a radiolabelled 75 g glucose drink. BP was assessed with an automated device, GE by scintigraphy and SMA flow by ultrasonography. Blood glucose and serum insulin were measured. GLP-1 attenuated the fall in diastolic BP after the glucose drink in older individuals (p < 0.05) and attenuated the fall in systolic and diastolic BP in patients with type 2 diabetes (p < 0.05). GE was faster in patients with type 2 diabetes than in healthy individuals (p < 0.05). In both groups, individuals had slower GE (p < 0.001), decreased SMA flow (p < 0.05) and a lower degree of glycaemia (p < 0.001) when receiving GLP-1. Intravenous GLP-1 attenuates the hypotensive response to orally administered glucose and decreases SMA flow, probably by slowing GE. GLP-1 and 'short-acting' GLP-1 agonists may be useful in the management of postprandial hypotension.
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
3-O-Methyl-D-glucopyranose, ≥98% (GC)