Merck
  • Resveratrol suppresses vascular endothelial growth factor secretion via inhibition of CXC-chemokine receptor 4 expression in ARPE-19 cells.
Resveratrol suppresses vascular endothelial growth factor secretion via inhibition of CXC-chemokine receptor 4 expression in ARPE-19 cells.
Molecular medicine reports (2015-03-31)
Hyemin Seong, Jinhyun Ryu, Joo Yeon Jeong, In Young Chung, Yong-Seop Han, Soo Hyun Hwang, Jong Moon Park, Sang Soo Kang, Seong Wook Seo
ABSTRACT
The present study characterizes the effects of resveratrol (Res) on vascular endothelial growth factor (VEGF) secretion in retinal pigment epithelial (RPE) cells. ARPE-19 cells were treated with CoCl2, a hypoxia mimetic agent. CoCl2 treatment increased protein levels of hypoxia inducible factor-1α (HIF-1α) and CXC-chemokine receptor 4 (CXCR4), and secretion of VEGF. To confirm the effects of Res on VEGF secretion, the human umbilical vein endothelial cell tube formation assay was performed with conditioned medium from Res-treated ARPE-19 cells. The well-known antioxidant Res effectively blocked these effects and reduced phosphorylation of nuclear factor (NF)-κB, an upstream activator of CXCR4. Furthermore, Res also suppressed VEGF secretion induced by SDF-1, a ligand of CXCR4. Conditioned medium from Res-treated ARPE-19 cells clearly suppressed tube formation compared with hypoxia-treated conditioned medium. The results demonstrated that Res inhibited the hypoxia mimetic CoCl2-induced expression of VEGF in ARPE-19 cells. Res suppressed CXCR4 expression through decreased phosphorylation of NF-κB, resulting in downregulation of VEGF secretion.
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%