Merck
  • Online open-tubular fractionation scheme coupled with push-pull perfusion sampling for profiling extravasation of gold nanoparticles in a mouse tumor model.
Online open-tubular fractionation scheme coupled with push-pull perfusion sampling for profiling extravasation of gold nanoparticles in a mouse tumor model.
Journal of chromatography. A (2015-05-26)
Cheng-Kuan Su, Po-Jen Tseng, Meng-Han Lin, Hsien-Ting Chiu, Andrea del Vall, Yu-Fen Huang, Yuh-Chang Sun
ABSTRACT
The extravasation of administered nano-drug carriers is a critical process for determining their distributions in target and non-target organs, as well as their pharmaceutical efficacies and side effects. To evaluate the extravasation behavior of gold nanoparticles (AuNPs), currently the most popular drug delivery system, in a mouse tumor model, in this study we employed push-pull perfusion (PPP) as a means of continuously sampling tumor extracellular AuNPs. To facilitate quantification of the extravasated AuNPs through inductively coupled plasma mass spectrometry, we also developed a novel online open-tubular fractionation scheme to allow interference-free determination of the sampled extracellular AuNPs from the coexisting biological matrix. After optimizing the flow-through volume and flow rate of this proposed fractionation scheme, we found that (i) the system's temporal resolution was 7.5h(-1), (ii) the stability presented by the coefficient of variation was less than 10% (6-h continuous measurement), and (iii) the detection limits for the administered AuNPs were in the range 0.057-0.068μgL(-1). Following an intravenous dosage of AuNPs (0.3mgkg(-1) body weight), in vivo acquired profiles indicated that the pegylated AuNPs (PEG-AuNPs) had greater tendency toward extravasating into the tumor extracellular space. We also observed that the accumulation of nanoparticles in the whole tumor tissues was higher for PEG-AuNPs than for non-pegylated ones. Overall, pegylation appears to promote the extravasation and accumulation of AuNPs for nano-drug delivery applications.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
SKU
Pack Size
Availability
Price
Quantity
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Nitric acid, 70%, purified by redistillation, ≥99.999% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Nitric acid, ACS reagent, ≥90.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Phosphate buffered saline, powder, pH 7.4, for preparing 5 L solutions