Merck
  • Alkaline comet assay in liver and stomach, and micronucleus assay in bone marrow, from rats treated with 2-acetylaminofluorene, azidothymidine, cisplatin, or isobutyraldehyde.
Alkaline comet assay in liver and stomach, and micronucleus assay in bone marrow, from rats treated with 2-acetylaminofluorene, azidothymidine, cisplatin, or isobutyraldehyde.
Mutation research. Genetic toxicology and environmental mutagenesis (2015-07-28)
A R Kraynak, J E Barnum, C L Cunningham, A Ng, B A Ykoruk, B Bennet, D Stoffregen, M Merschman, E Freeland, S M Galloway
ABSTRACT
As part of the Japanese Center for the Validation of Alternative Methods (JaCVAM) initiative international validation study of the in vivo rat alkaline comet assay (comet assay), we examined the ability of the assay to determine the genotoxicity of 2-acetylaminofluorene (AAF), azidothymidine (AZT), cisplatin (CPN), and isobutyraldehyde (IBA) in liver and glandular stomach of male Sprague-Dawley rats. Rats were given oral doses of test compound or control once daily for three days. High dose levels were approximately maximum tolerated doses and were based on preliminary range-finding studies. Tissues were harvested 3h after the final dose (48h after the initial dose). A bone marrow micronucleus assay (MN) was also conducted on the rats treated with AZT, CPN, and IBA. Acute toxic effects of treatment were determined primarily through histomorphologic analysis of liver and stomach but also by body weight and serum liver enzyme changes. The comet assay was conducted on fresh tissue preparations but frozen samples from two studies were also assayed. Statistically significant dose-related differences in comet % DNA in tail were found in liver and stomach for the genotoxin AZT and in liver for the genotoxin CPN, but not in liver or stomach for the non-genotoxin IBA. Statistically significant differences in % DNA in tail were measured in liver for the low and mid dose of the genotoxin AAF, but not the high dose. The comet assays of frozen liver suspensions from CPN- and AAF-treated rats yielded comparable results to the assays of fresh preparations. There were no indications of significant toxicity induced by any treatment. The micronucleus assay was positive for CPN and AZT and negative for IBA. In conclusion, the in vivo comet assay is capable of detecting genotoxic effects of a variety of chemicals and may fill an important role in the genotoxicity test battery.
Product Description
SAFC
Sodium chloride solution, 5 M
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride solution, 0.85%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Trizma® base, ≥99.9% (titration), crystalline
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Tromethamine, meets USP testing specifications
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride, tablet
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Trizma® base, BioXtra, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Trizma® base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acridine Orange hydrochloride solution, 10 mg/mL in H2O, ≥95.0% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride solution, 5 M
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Trizma® base, puriss. p.a., ≥99.7% (T)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Trizma® base, ≥99.0% (T)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Trizma® base, BioUltra, for molecular biology, ≥99.8% (T)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ACS reagent, ≥99.8%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis