Merck
  • Two-dimensional paper chromatography-based fluorescent immunosensor for detecting acute myocardial infarction markers.
Two-dimensional paper chromatography-based fluorescent immunosensor for detecting acute myocardial infarction markers.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-08-05)
Jung-Hwan Cho, Min-Ha Kim, Rak-Sun Mok, Jin-Woo Jeon, Guei-Sam Lim, Chan-Young Chai, Se-Hwan Paek
ABSTRACT
A novel washing scheme following antigen-antibody reactions with analyte was used during construction of a fluorescent immunosensor to resolve the background problem in the lateral flow assay with human serum. An immuno-membrane strip was devised to simultaneously measure cardiac troponin I (cTnI), creatinine kinase-MB isoform (CK-MB), and myoglobin to diagnose acute myocardial infarction. This strip was then installed within a cartridge containing a built-in washing solution tank, which was used to supply the solution across the signal generation pad of the strip after the immune reactions. Such cross-flow washing was initiated by onset-signaling from the internal control and began to run automatically upon sample addition. Under optimal conditions, the immunosensor displayed a stably suppressed background baseline, enabling us to attain a low detection limit for cTnI (0.05 ng/mL) as well as favorable reproducibility for repetitive measurements (relative standard deviation <10%). No interference was observed among the different complex formations at the respective analyte sites, and no artifacts were caused by sample matrices. We tested the performance relationship with the Pathfast reference system for positive serum samples (36 for cTnI, 58 for CK-MB, and 17 for myoglobin), and the correlation coefficients were >0.98. This result suggests that the new immunosensor system based on two-dimensional chromatography can be used for clinical testing.
Product Description
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium phosphate dibasic solution, BioUltra, 0.5 M in H2O
Sigma-Aldrich
Sodium phosphate monobasic-16O4, 99.9 atom % 16O
Sigma-Aldrich
Sodium chloride solution, 0.85%
Supelco
Phosphate Standard for IC, TraceCERT®, 1000 mg/L phosphate in water (nominal concentration)
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium phosphate monobasic solution, BioUltra, 5 M in H2O
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium phosphate monobasic, purum p.a., anhydrous, ≥99.0% (T)
Millipore
Sucrose, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sodium phosphate dibasic, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Sodium phosphate dibasic, purum p.a., anhydrous, ≥98.0% (T)