Merck
  • miR-184 regulates ezrin, LAMP-1 expression, affects phagocytosis in human retinal pigment epithelium and is downregulated in age-related macular degeneration.
miR-184 regulates ezrin, LAMP-1 expression, affects phagocytosis in human retinal pigment epithelium and is downregulated in age-related macular degeneration.
The FEBS journal (2014-09-25)
Najiba Murad, Maria Kokkinaki, Nishantha Gunawardena, Mia S Gunawan, Yetrib Hathout, Karolina J Janczura, Alexander C Theos, Nady Golestaneh
ABSTRACT
MicroRNA 184 (miR-184) is known to play a key role in neurological development and apoptosis and is highly expressed in mouse brain, mouse corneal epithelium, zebrafish lens and human retinal pigment epithelium (RPE). However, the role of miR-184 in RPE is largely unknown. We investigated the role of miR-184 in RPE and its possible implication in age-related macular degeneration (AMD). Proteomic analysis identified the ezrin (EZR) gene as a target of miR-184 in human RPE. EZR is a membrane cytoskeleton crosslinker that is also known to bind to lysosomal-associated membrane protein 1 (LAMP-1) during the formation of phagocytic vacuoles. In adult retinal pigment epithelium 19 (ARPE19) cells, inhibition of miR-184 resulted in upregulation of EZR mRNA and EZR protein, and induced downregulation of LAMP-1. The inhibition of miR-184 decreased EZR-bound LAMP-1 protein levels and affected phagocytic activity in ARPE19 cells. In primary culture of human RPE isolated from eyes of AMD donors (AMD RPE), miR-184 was significantly downregulated compared with control (normal) RPE. Downregulation of miR-184 was consistent with significantly lower levels of LAMP-1 protein in AMD RPE, and overexpression of MIR-184 in AMD RPE was able to rescue LAMP-1 protein expression to normal levels. Altogether, these observations suggest a novel role for miR-184 in RPE health and support a model proposing that downregulation of miR-184 expression during aging may result in dysregulation of RPE function, contributing to retinal degeneration.
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Sodium Fluoride Solution
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture