Merck
  • pH-sensitive stearoyl-PEG-poly(methacryloyl sulfadimethoxine) decorated liposomes for the delivery of gemcitabine to cancer cells.
pH-sensitive stearoyl-PEG-poly(methacryloyl sulfadimethoxine) decorated liposomes for the delivery of gemcitabine to cancer cells.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V (2014-08-27)
Sara Bersani, Marian Vila-Caballer, Chiara Brazzale, Michela Barattin, Stefano Salmaso
ABSTRACT
Novel, acid-sensitive liposomes that respond to physiopathological pH for tumour targeting applications were obtained by surface decoration with 1,2-distearoyl-sn-glycero-3-phosphoethanolamine-N-[methoxy(polyethylene glycol)] (mPEG-DSPE) and stearoyl-poly(ethylene glycol)-poly(methacryloyl sulfadimethoxine) copolymer (stearoyl-PEG-polySDM). The pH-sensitive stearoyl-PEG-polySDM copolymer contained an average of seven methacryloyl sulfadimethoxines per molecule and was found to possess an apparent pKa of 7.2. Preliminary cloud point studies showed that the hydrophilic/hydrophobic copolymer conversion occurred at pH 7.0. The copolymer was soluble above pH 7.0 and underwent aggregation at lower pH. Liposome formulations were prepared with 0.2:0.6:100, 0.5:1.5:100 and 1:3:100 mPEG-DSPE/stearoyl-PEG-polySDM/lipids molar ratios. All of the liposome formulations were stable at pH 7.4, even in the presence of foetal bovine serum, but they underwent rapid size increase at pH 6.5. TEM analysis showed that, at pH 6.5, the formulations coated with a stearoyl-PEG-polySDM/lipids molar ratio greater than 1:100 underwent aggregation. At pH 7.4, the liposomes showed negative zeta potential that significantly decreased after incubation at pH 6.5. Cell-culture studies indicated that the liposomes were not toxic up to 10mg/mL. Fluorescence spectroscopy, cytofluorimetry and confocal microscopy showed that at pH 6.5, the incubation of MCF-7 tumour cells with fluorescein-labelled 1:3:100 mPEG-DSPE/stearoyl-PEG-polySDM/lipids molar ratio liposomes resulted in time-dependent cell association, while at pH 7.4 the cell interaction was significantly lower. The same pH-responsive liposome formulation loaded with gemcitabine (98.2±4.7nmol gemcitabine/lipid μmol loading capacity) was stable at pH 7.4 for several hours, while at pH 6.5 it rapidly aggregated. At pH 6.5, these liposomes displayed higher cytotoxicity than at pH 7.4 or compared to non-responsive control liposomes at both incubation pH. Notably, treatment with free gemcitabine did not yield cytotoxic effects, indicating that the carrier can efficiently deliver the anticancer drug to the cytosolic compartment.
Product Description
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
O-Phosphorylethanolamine
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%