Merck
  • S100A4 is upregulated in proliferative diabetic retinopathy and correlates with markers of angiogenesis and fibrogenesis.
S100A4 is upregulated in proliferative diabetic retinopathy and correlates with markers of angiogenesis and fibrogenesis.
Molecular vision (2014-09-26)
Ahmed M Abu El-Asrar, Mohd Imtiaz Nawaz, Gert De Hertogh, Kaiser Alam, Mohammad Mairaj Siddiquei, Kathleen Van den Eynde, Ahmed Mousa, Ghulam Mohammad, Karel Geboes, Ghislain Opdenakker
ABSTRACT
The calcium-binding protein S100A4 is implicated in cancer cell invasion and metastasis, the stimulation of angiogenesis, the progression of fibrosis, and inflammatory disorders. We investigated the expression of S100A4 and correlated it with clinical disease activity as well as with the levels of osteopontin (OPN), soluble syndecan-1, and vascular endothelial growth factor (VEGF) in proliferative diabetic retinopathy (PDR). To reinforce the findings at the functional level, we examined the expressions of S100A4 and OPN in the retinas of diabetic rats and in human retinal microvascular endothelial cells (HRMECs) following exposure to VEGF and the proinflammatory cytokine tumor necrosis factor-α (TNF-α). Vitreous samples from 30 PDR and 30 nondiabetic patients, epiretinal membranes from 14 patients with PDR, the retinas of rats, and HRMECs were studied by enzyme-linked immunosorbent assay (ELISA), immunohistochemistry, western blot analysis, and co-immunoprecipitation. ELISA revealed a significant increase in the expressions of S100A4, OPN, soluble syndecan-1, and VEGF in vitreous samples from PDR patients compared to nondiabetic controls (p = 0.001; <0.001; <0.001; <0.001, respectively). Significant positive correlations were found between the levels of S100A4, OPN (r = 0.52, p = <0.001), soluble syndecan-1 (r = 0.37, p = 0.012), and VEGF (r = 0.29, p = 0.044). In epiretinal membranes, S100A4 was expressed in the vascular endothelial cells and stromal CD45-expressing leukocytes. A significant positive correlation was detected between the number of blood vessels expressing CD31 and the number of stromal cells expressing S100A4 (r = 0.77, p = 0.001). Western blot analysis revealed a significant increase in the expressions of S100A4 and both intact and cleaved OPN in vitreous samples from PDR patients compared to nondiabetic controls, as well as in the retinas of diabetic rats. Co-immunoprecipitation studies revealed a positive interaction between S100A4 and the receptor for advanced glycation end products (RAGE) in the retinas of diabetic rats. TNF-α-but not VEGF-induced the upregulations of S100A4 and both intact and cleaved OPN in HRMECs. S100A4 represents a valuable vitreous marker molecule in the pathogenesis of PDR and might become a new target for the treatment of PDR.
Sigma-Aldrich
Citric acid trisodium salt, anhydrous, ≥98% (GC)
Supelco
Sulfuric acid, for the determination of nitrogen, ≥97.0%
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Citrate Concentrated Solution, BioUltra, 1 M in H2O
Sigma-Aldrich
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
Sigma-Aldrich
Sulfuric acid, ACS reagent, 95.0-98.0%
Supelco
Sodium Citrate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sulfuric acid, puriss., meets analytical specification of Ph. Eur., BP, 95-97%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Luminol, 97%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Luminol, ≥97% (HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Supelco
Sulfuric acid concentrate, 0.1 M H2SO4 in water (0.2N), eluent concentrate for IC
Sigma-Aldrich
Human Angiostatin ELISA Kit, serum, plasma, cell culture supernatant and urine