Merck
  • Simultaneous determination of sixteen amide fungicides in vegetables and fruits by dispersive solid phase extraction and liquid chromatography-tandem mass spectrometry.
Simultaneous determination of sixteen amide fungicides in vegetables and fruits by dispersive solid phase extraction and liquid chromatography-tandem mass spectrometry.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2015-03-24)
Yin-Liang Wu, Ruo-Xia Chen, Yong Zhu, Jian Zhao, Ting Yang
ABSTRACT
A modified quick, easy, cheap, effective, rugged, and safe (QuEChERS) method using multi-walled carbon nanotubes (MWCNTs) as a reversed-dispersive solid phase extraction (r-dSPE) material combined with ultra-high liquid chromatography tandem mass spectrometry (UHPLC-MS/MS) was developed for the simultaneous determination of 16 novel amide fungicides in vegetables and fruits. After extraction with acetonitrile, a dSPE cleanup procedure, which was developed after the optimization of the type and amount of MWCNTs, the pH value of the extract, the extraction time for MWCNTs, and the type of eluent with MWCNTs material, was conducted. The determination of the target compounds was conducted in less than 7.0 min while the specificity is ensured through the MRM acquisition mode. The linearity of the analytical response across the studied range of concentrations (0.25-500 μg/L) was excellent, obtaining correlation coefficients higher than 0.997. The samples were quantified with the matrix matched standard solutions. The average recoveries in cabbage, celery, strawberry, and grape at three spiked levels (0.01, 0.5, and 5.0mg/kg) were ranged from 72.4 to 98.5% with all RSDs lower than 10%. The limits of detection were below 0.003 mg/kg and the limits of quantification did not exceed 0.01 mg/kg in all matrices. The method demonstrated to be suitable for the simultaneous determination of 16 novel amide fungicides in vegetables and fruits.
Product Description
Supelco
Silthiofam, PESTANAL®, analytical standard
Supelco
Zoxamide, PESTANAL®, analytical standard
Supelco
Carpropamid, PESTANAL®, analytical standard
Supelco
Fluopicolide, PESTANAL®, analytical standard
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Supelco
Fenhexamid, PESTANAL®, analytical standard
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Methanol solution, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Thifluzamide, PESTANAL®, analytical standard
Supelco
Cyflufenamid, PESTANAL®, analytical standard
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Methanol solution, (Methanol:Dimethyl sulfoxide 1:1 (v/v))
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, 10 M in H2O