Skip to Content
Merck
All Photos(1)

Key Documents

41132

Supelco

Fluopicolide

PESTANAL®, analytical standard

Synonym(s):

2,6-Dichloro-N-[3-chloro-5-(trifluoromethyl)-2-pyridylmethyl]benzamide

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€203.00

€203.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€203.00

About This Item

Empirical Formula (Hill Notation):
C14H8Cl3F3N2O
CAS Number:
Molecular Weight:
383.58
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€203.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

FC(F)(F)c1cnc(CNC(=O)c2c(Cl)cccc2Cl)c(Cl)c1

InChI

1S/C14H8Cl3F3N2O/c15-8-2-1-3-9(16)12(8)13(23)22-6-11-10(17)4-7(5-21-11)14(18,19)20/h1-5H,6H2,(H,22,23)

InChI key

GBOYJIHYACSLGN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Melatonin Attenuates Potato Late Blight by Disrupting Cell Growth, Stress Tolerance, Fungicide Susceptibility and Homeostasis of Gene Expression in
Shumin Zhang et al.
Frontiers in plant science, 8, 1993-1993 (2017-12-07)
Jie Wu et al.
Frontiers in microbiology, 11, 185-185 (2020-03-21)
Sensitivities of Phytophthora capsici to fluopicolide were investigated in vitro, with results showing that fluopicolide had strong inhibitory activities on each development stage of P. capsici, in particular on the motility of the zoospore. The potential resistance risk for fluopicolide
Juliana González-Tobón et al.
Plant disease, 104(1), 211-221 (2019-11-26)
Phytophthora infestans is the causal agent of late blight disease of potatoes and tomatoes. This disease causes devastating economic losses each year, and control is mainly achieved by the use of fungicides. Unfortunately, populations of P. infestans resistant to fungicides
Manuel PatiÑo et al.
Journal of food protection, 83(10), 1726-1737 (2020-09-25)
In this study, chemical and microbiological contaminants were detected in fresh fruits and vegetables in peasant markets in the Department of Cundinamarca to assess food safety. At least one pesticide was detected in 63% of the samples evaluated (n =
Zhiwen Wang et al.
Pest management science, 76(7), 2525-2536 (2020-02-23)
Phytophthora capsici is a destructive plant oomycete pathogen that could lead to devastating losses in food production. Fungicide application is the main way to control plant disease caused by P. capsici. SYP-14288, a novel fungicide with a unique mode of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service