Merck
  • Shell-programmed Au nanoparticle heterodimers with customized chiroptical activity.
Shell-programmed Au nanoparticle heterodimers with customized chiroptical activity.
Small (Weinheim an der Bergstrasse, Germany) (2014-08-20)
Yuan Zhao, Liguang Xu, Wei Ma, Liqiang Liu, Libing Wang, Hua Kuang, Chuanlai Xu
ABSTRACT
Chiral plasmonic assemblies with strong and tunable chiroptical activity are emerging materials yet challenging to fabricate. Moreover, shell-programmed chiroptical regulation is really rare. Here, the chiroptical activity of core-shell (CS) nanoparticles (NPs) heterodimers (HDs) with different types and thicknesses of the shell but featuring the same gap was exploited. It was found that the type of shell guided the position of the chiral peaks, and the shell thickness tuned the intensity but also moderately affected the wavelength shift at invariable interparticle distance. Shell deposition intensified the hot-spot chirality, and evidently guided the enantiomorphous chiral configuration, resulting in a startlingly intense, asymmetric, dipolar coupling strength. The magnitude of the chiroptical activity showed an 8-10 fold enhancement with a maximum anisotropy factor (g-factor) of 1.5 × 10(-2) . Shell-driven chiroptical regulation opens new avenues to feasibly fabricate chiroptically active materials with desired chiroptical response for the development of switchable recognition units for sensitive and various target detections.
Sigma-Aldrich
Hydroxylamine hydrochloride, 99.995% trace metals basis
Sigma-Aldrich
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
Sigma-Aldrich
Citrate Concentrated Solution, BioUltra, for molecular biology, 1 M in H2O
Sigma-Aldrich
Hydroxylamine hydrochloride, 99.999% trace metals basis
Supelco
Sodium Citrate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Hydroxylamine hydrochloride, ReagentPlus®, 99%
Sigma-Aldrich
Hydroxylamine hydrochloride, ACS reagent, 98.0%
Lysine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Hydroxylamine hydrochloride, puriss. p.a., ACS reagent, ≥99.0% (RT)
Supelco
L-Lysine monohydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Supelco
L-Lysine hydrochloride solution, 100 mM amino acid in 0.1 M HCl, analytical standard
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Supelco
L-Lysine monohydrochloride, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland