Merck
  • Polymorphisms of the thiopurine S-methyltransferase gene among the Libyan population.
Polymorphisms of the thiopurine S-methyltransferase gene among the Libyan population.
The Libyan journal of medicine (2015-03-31)
Hamza Ben Zeglam, Abdrazak Benhamer, Adel Aboud, Haitem Rtemi, Meftah Mattardi, Saleh Suleiman Saleh, Abdullah Bashein, Nabil Enattah
ABSTRACT
Thiopurine S-methyltransferase (TPMT) is a cytosolic enzyme that catalyses the S-methylation of 6-mercaptopurine and azathioprine. Low activity phenotypes are correlated with polymorphism in the TPMT gene. Patients with low or undetectable TMPT activity could develop severe myelosuppression when they are treated with standard doses of thiopurine drugs. Since ethnic differences in the TPMT gene polymorphism have been demonstrated worldwide, assessing it in the Libyan population is worthwhile. We investigated TPMT gene polymorphism in a total of 246 Libyan healthy adult blood donors from three different Libyan regions (Tripoli, Yefren, and Tawargha) and 50 children with acute lymphoblastic leukaemia (ALL). We used polymerase chain reaction restriction length polymorphism (PCR-RFLP) and allele-specific PCR-based assays to analyse the TPMT gene for the variants *2 c.238 G>C, *3A (c.460 G>A and c.719 A>G), *3B (c.460 G>A), and *3C (c.719 A>G). Our results show that the TPMT variants associated with low enzymatic activity were detected in 3.25% (8 in 246) of adult Libyan individuals and the frequency of total mutant alleles was 1.63%. Heterozygous genotypes were TPMT*3A in three subjects (0.61%) and TPMT*3C in five subjects (1.02%). No TPMT*2 and TPMT*3B allelic variants and no homozygous or compound heterozygous mutant alleles were detected. The normal allele (wild-type) was found in 98.4% of the adult individuals studied. No mutant alleles were detected among the 50 children who had ALL. We report on the presence of the TPMT*3C and *3A mutant alleles in the Libyan population. Therefore, monitoring the patients to be treated with doses of thiopurine drugs for TPMT variants is worthwhile to avoid the development of severe myelosuppression.
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Supelco
DL-Dithiothreitol solution, 1 M in H2O
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis